Stampa…

Piombo (II) cloruro 99%, per analisi

Fornitore: Thermo Fisher Scientific
Pericolo

Synonyms: Lead dichloride

12349.18 12349.36 12349.A3
12349.18EA 22.4 EUR
12349.18 12349.36 12349.A3
Piombo (II) cloruro 99%, per analisi
Piombo (II) cloruro

Purity: 99% CAS No.: 7758-95-4 Molecular Formula: PbCl2 Formula Weight: 278.10 Physical Form: Crystalline Powder Density: 5.85 Melting Point: 501° Boiling Point: 951° Solubility: Soluble in hot water, alkali hydroxides, NH4Cl solution. Insoluble in cold water and alcohol Merck Reference: 14,5404 UN No.: 2291 MDL No.: MFCD00011157


Analytical reagent, preparation of lead salts, as solder and flux

Formula: Cl₂Pb
Peso molecolare: 278.11 g/mol
Punto di ebollizione: 951 °C
Punto di fusione: 501 °C
Densità: 5.85
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00011157
CAS Number: 7758-95-4
EINECS: 231-845-5
UN: 2291
ADR: 6.1 III
Merck Index: 14,5404

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR