Stampa…

Bario fluoruro ≥99%

Fornitore: Thermo Fisher Scientific
Attenzione

12338.14 12338.30 12338.A1
12338.14EA 21.3 EUR
12338.14 12338.30 12338.A1
Bario fluoruro ≥99%
Bario fluoruro

Purity: 99% CAS No.: 7787-32-8 Molecular Formula: BaF2 Formula Weight: 175.36 Physical Form: -40 Mesh Powder Density: 4.83 Refractive Index: 1.4741 Odour: Odourless Melting Point: 1353° Boiling Point: 2260° Solubility: Soluble in HCl, HNO3, HF, and aqueous solutions of ammonium chloride Merck Reference: 14,974 UN No.: 1564 MDL No.: MFCD00003450


Ceramic flux, glass manufacture, electronics, and dry-film lubricants

Formula: BaF₂
Punto di ebollizione: 2260 °C
Punto di fusione: 1353 °C
Densità: 4,83
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00003450
CAS Number: 7787-32-8
EINECS: 232-108-0
UN: 1564
ADR: 6.1,III
Merck Index: 14,974

Visualizza i codici

Specifiche

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR