Stampa…

Magnesio cloruro, anidro 99%

Fornitore: Thermo Fisher Scientific
Attenzione

12315.A1 12315.A7 12315.C4
12315.A1EA 59.5 EUR
12315.A1 12315.A7 12315.C4
Magnesio cloruro, anidro 99%
Magnesio cloruro

Purity: 99% CAS No.: 7786-30-3 Molecular Formula: MgCl2 Formula Weight: 95.22 Physical Form: Crystalline Density: 2.32 Appearance: White to yellow Odour: Odourless Melting Point: 714° Solubility: Soluble in water with evolution of heat Merck Reference: 14,5662 MDL No.: MFCD00011106


Catalyst, flocculating agent, various magnesium cements, ceramics, analytical reagent

Formula: MgCl₂
Punto di ebollizione: 1412 °C (1013 hPa)
Punto di fusione: 714 °C
Densità: 2,32
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00011106
CAS Number: 7786-30-3
EINECS: 232-094-6
Merck Index: 14,5662

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR