Stampa…

Calcio cloruro esaidrato 97-103%, meets analytical specification of Ph. Eur., E509 (max. 0,0001% Al), Fluka™

Fornitore: Honeywell Chemicals
Attenzione

12074-1KG 12074-2.5KG 12074-6X2.5KG
12074-1KGEA 70.49 EUR
12074-1KG 12074-2.5KG 12074-6X2.5KG
Calcio cloruro esaidrato 97-103%, meets analytical specification of Ph. Eur., E509 (max. 0,0001% Al), Fluka™
Calcio cloruro esaidrato

  • Contains ≤0.0001% Al.

Formula: CaCl₂·6H₂O
Peso molecolare: 219.08 g/mol
Punto di ebollizione: 1.900 °C (1.013 hPa)
Punto di fusione: 29 °C
Densità: 1.680 g/cm3 (20 °C)
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD00149616
CAS Number: 7774-34-7
Merck Index: 13,01660

Visualizza i codici

Risultati del test di specificazione

Transition Temp solidification point 28-30 °C
Identity complying
Assay 97.0 - 103.0 %
Solidification range 28 - 30 °C
Acid. or alk. react. impurities complying
Aluminium (Al) Max. 1 ppm
Arsenic (As) Max. 1 ppm
Barium (Ba) complying
Iron (Fe) Max. 5 ppm
Mercury (Hg) Max. 1 ppm
Lead (Pb) Max. 5 ppm
Heavy metals (as Pb) Max. 10 ppm
Magnesium and alkali metals Max. 0.3 %
Fluoride (F) Max. 40 ppm
Sulfate (SO₄) Max. 100 ppm
Residual solvents complying
Appearance of the solution complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR