Stampa…

Dipotassium hexachloropalladate (≥26,3% Pd)

Fornitore: Thermo Fisher Scientific
Pericolo

Synonyms: Potassium hexachloropalladate(IV)

11884.03 11884.06 11884.14
11884.03EA 86 EUR
11884.03 11884.06 11884.14
Dipotassium hexachloropalladate (≥26,3% Pd)
Dipotassium hexachloropalladate

CAS No.: 16919-73-6 Molecular Formula: K2PdCl6 Formula Weight: 397.32 Physical Form: Crystalline Density: 2.738 Sensitivity: Hygroscopic Appearance: Red Odour: Odourless Melting Point: dec. UN No.: 3077 MDL No.: MFCD00043081

Formula: Cl₆K₂Pd
Peso molecolare: 397.33 g/mol
Punto di fusione: Dec.
Densità: 2.738
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00043081
CAS Number: 16919-73-6
EINECS: 240-974-6
ADR: 9 III

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR