Stampa…

Bismuth triacetate ≥99.999% (metals basis)

Fornitore: Thermo Fisher Scientific

Synonyms: Acetic acid bismuth (III) salt, Bismuth (III) acetate, Bismuth(III) acetate

11846.09 11846.18
11846.09EA 148 EUR
11846.09 11846.18
Bismuth triacetate ≥99.999% (metals basis)
Bismuth triacetate

Purity: 99.999% CAS No.: 22306-37-2 Molecular Formula: Bi(OOCCH3)3 Formula Weight: 386.12 Physical Form: Crystalline Sensitivity: Hygroscopic MDL No.: MFCD00015629

Formula: (H₃CCOO)₃Bi
Punto di fusione: Dec.
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00015629
CAS Number: 22306-37-2
EINECS: 244-904-5

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR