Stampa…
Electrical/pneumatic lockout, large
Fornitore: Brady
800126
118-0536EA 52.14 EUR
118-0536
Electrical/pneumatic lockout, large
Chiusure di sicurezza
This pneumatic lockout accommodates a large variety of electrical plugs up to 89 mm diameter and 127 mm in length as well as male air hose connectors.
  • Made of PS
  • Holes are designed to fit 'necked down' pneumatic male fittings commonly attached to compressed air hoses
  • Suitable for 110 V, 220 V NS 550 V plugs
  • Four padlocks can be applied
Max. shackle diameter: 7,00 mm
Min. shackle clearance: 20,00 mm
Plug diameter: 89,00 mm
Plug length: 127,00 mm