Stampa…
Cleanroom wipes, presaturated, PROSAT® Sterile™ Theta™
Fornitore: CONTEC
PSC20005-BPR
115-2782EA 917 EUR
115-2782
Cleanroom wipes, presaturated, PROSAT® Sterile™ Theta™
Panni Panni per camera bianca Preimbevuti
These PROSAT® Sterile™ Theta™ wipes are manufactured from a lightweight hydroentangled cellulose/polyester and saturated with a solution of 70% USP grade IPA and 30% deionized water.
  • Suitable for Grade A/B or ISO Class 5-8 environments
  • Validated sterile per gamma irradiation
  • Hydroentangled cellulose/polyester fabric saturated with 70% USP grade IPA and 30% DI water
  • Nonwoven material which is low in particles and fibres making an excellent general purpose wipe
  • Highly sorbent with good wet strength
  • Presaturated wipe reduces VOC emissions and solvent use