Stampa…

Cerio (III) nitrato esaidrato ≥99.5% (REO, rare earth oxide basis), REacton®

Fornitore: Thermo Fisher Scientific
Pericolo

Synonyms: Cerous chloride hexahydrate, Cerium (III) nitrate hexahydrate

11329.09 11329.22 11329.36 11329.A1
11329.09EA 47.1 EUR
11329.09 11329.22 11329.36 11329.A1
Cerio (III) nitrato esaidrato ≥99.5% (REO, rare earth oxide basis), REacton®
Cerio (III) nitrato esaidrato

Purity: 99.5% CAS No.: 10294-41-4 Molecular Formula: CeN3O9.6H2O Formula Weight: 434.23 (326.13anhy) Physical Form: Crystalline Aggregates Sensitivity: Hygroscopic pH: 3 Solubility: Soluble in water, alcohol, acetone Merck Reference: 14,2000 UN No.: 1477 MDL No.: MFCD00149631


Separation of cerium from other rare earths, catalyst for hydrolysis of phosporic acid esters

Formula: Ce(NO₃)₃·6H₂O
Peso molecolare: 434.22 g/mol
Punto di ebollizione: 200
Punto di fusione: 39
Densità: 2.377 g/cm³ (20 °C)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00149631
CAS Number: 10294-41-4
EINECS: 233-297-2
UN: 1477
ADR: 5.1 III
Merck Index: 14,2000

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR