Stampa…
VWR®, Copriscarpe, LDPE/LLDPE
Fornitore: VWR Collection
113-8281EA 9.6 EUR
113-8281
VWR®, Copriscarpe, LDPE/LLDPE
Indumenti usa e getta
Shoe covers made from CPE, a mixture of 65% Low Density PE (LDPE) and 35% Linear Low Density PE (LLDPE).
  • Repellent against moisture and dirt
  • No seam under the foot
L×H: 410×150 mm
* One size fits all
Visualizza i codici
Specifiche
Environmentally Preferable
Genere
Applicazioni per abiti Laboratory