Stampa…
Cleanroom face masks, BioClean Microflow™
Fornitore: Nitritex
BFV05
113-8158EA 892 EUR
113-8158
Cleanroom face masks, BioClean Microflow™
Maschere Maschere per cleanroom
Non sterile soft face veil with headband and nosepiece manufactured using ultrasonic bonding.
  • Veil made from low linting thermabond non woven material
  • White hydroentangled polyester binding and tubular polyester head loop
  • Fully enclosed, flexible, malleable aluminium nosepiece coated with plastic
  • Processed in a cleanroom environment, contains no latex
Suitable for use in class 10, ISO Class 4, M2.5 cleanrooms.
Certificazioni: Manufactured in a facility holding ISO 9001:2008. Processed in an NEBB certified ISO class 4 cleanroom environment. SamplingMore Product Information