Stampa…
Self-retracting lifelines, Miller® Falcon™
Fornitore: Honeywell Safety
1016796
113-1279EA 718 EUR
113-1279
Self-retracting lifelines, Miller® Falcon™
Protezione anticaduta Dispositivi di salvataggio
This self-retracting lifeline is the ideal fall protection solution for construction, industry, warehouses, utilities, mining and confined space applications.
  • Quick-activating braking mechanism stops a fall within centimetres
  • Lightweight
  • No annual factory re-certification required: Reduced down time, lower cost of ownership and increased productivity
  • High-impact resistant polyamide housing, polyester webbing
Certificazioni: EN 360
Visualizza i codici
Specifiche
Modello Falcon™
Environmentally Preferable
Tipo di cavo Polyester webbing
Tipo di connettore Snap hook, aluminium
Tensione > 1500 daN
Temperatura d'uso −30...+50 °C
L×P×A alloggiamento 540×145×70 mm