Stampa…

Europio (III) ossido ≥99.99% (REO, rare earth oxide basis), REacton®

Fornitore: Thermo Fisher Scientific

11299.03 11299.06 11299.14 11299.22
11299.03EA 42.6 EUR
11299.03 11299.06 11299.14 11299.22
Europio (III) ossido ≥99.99% (REO, rare earth oxide basis), REacton®
Europio (III) ossido

Purity: 99.99% CAS No.: 1308-96-9 Molecular Formula: Eu2O3 Formula Weight: 351.92 Physical Form: Powder Sensitivity: Hygroscopic Merck Reference: 14,3904 MDL No.: MFCD00010997

Formula: Eu₂O₃
Punto di fusione: 2291 °C
Densità: 7,42 g/cm³ (20 °C)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00010997
CAS Number: 1308-96-9
EINECS: 215-165-6
Merck Index: 14,3904

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR