Stampa…

Acido carminico, Supelco®

Fornitore: Merck


MQ100

11298-25MG
11298-25MGEA 167 EUR
11298-25MG
Acido carminico, Supelco®
Carminic acid
Formula: C₂₂H₂₀O₁₃
MDL Number: MFCD00167028
CAS Number: 1260-17-9
EINECS: 215-023-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db47fc59d85298af14cae22fec0a8e3a5785b6dd&vid=ef2ed2531ef962c38c91c995d60d225aea094dad&prid=web&clid=SAPDAM

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR