Stampa…

Praseodymium(III) nitrate hexahydrate ≥99.99% (REO, rare earth oxide basis), REacton®

Fornitore: Thermo Fisher Scientific
Pericolo

11240.14 11240.22
11240.14EA 170 EUR
11240.14 11240.22
Praseodymium(III) nitrate hexahydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Praseodymium(III) nitrate hexahydrate

Purity: 99.99% CAS No.: 15878-77-0 Molecular Formula: Pr(NO3)3.6H2O Formula Weight: 435.01 (326.92anhy) Physical Form: Crystalline Aggregates Sensitivity: Hygroscopic UN No.: 1477 MDL No.: MFCD00149826

Formula: N₃O₉Pr·6H₂O
Punto di fusione: 56 °C
Densità: 2,233 g/cm³ (20 °C)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00149826
CAS Number: 15878-77-0
EINECS: 233-796-5
UN: 1477
ADR: 5.1,III

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR