Stampa…

Samario (III) ossido ≥99.99% (REO, rare earth oxide basis), REacton®

Fornitore: Thermo Fisher Scientific

11230.14 11230.22 11230.36
11230.14EA 155 EUR
11230.14 11230.22 11230.36
Samario (III) ossido ≥99.99% (REO, rare earth oxide basis), REacton®
Samario (III) ossido

Purity: 99.99% CAS No.: 12060-58-1 Molecular Formula: Sm2O3 Formula Weight: 348.70 Physical Form: Powder Refractive Index: 1.97 Melting Point: 2300° Merck Reference: 14,8347 MDL No.: MFCD00011237

Formula: O₃Sm₂
Punto di fusione: 2300 °C
Densità: 7,68 g/cm³ (20 °C)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00011237
CAS Number: 12060-58-1
EINECS: 235-043-6
Merck Index: 14,8347

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR