Stampa…
Face seal cleaning wipes for respirators, 105
Fornitore: 3M
105
111-1952EA 91.8 EUR
111-1952
Face seal cleaning wipes for respirators, 105
Panni Panni e asciugamani monouso Panni preimbevuti
The 105 face seal cleaner is an individually wrapped wipe for cleaning and freshening the face piece without damaging it.
  • Antiseptic
  • Will not dry out even after long periods of storage in warm conditions
  • Complies with current regulations requiring cleaning of non-disposable respiratory equipment after use
Confezione: Individually foil wrapped.