Stampa…

Magnesio fluoruro ≥99.9% (metals basis), Optical Grade Optical Grade

Fornitore: Thermo Fisher Scientific
Attenzione

11021.09 11021.18
11021.09EA 126 EUR
11021.09 11021.18
Magnesio fluoruro ≥99.9% (metals basis), Optical Grade Optical Grade
Magnesio fluoruro

Purity: 99.9% CAS No.: 7783-40-6 Molecular Formula: MgF2 Formula Weight: 62.30 Physical Form: Broken Crystals Density: 3.148 Refractive Index: 1.35-1.38 Appearance: White Odour: Odourless Melting Point: 1261° Merck Reference: 14,5665 MDL No.: MFCD00011108

Formula: MgF₂
Punto di ebollizione: 2260 °C (1013 hPa)
Punto di fusione: 1261 °C
Densità: 3,148
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00011108
CAS Number: 7783-40-6
EINECS: 231-995-1
Merck Index: 14,5665

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR