Stampa…

Itterbio (III) ossido ≥99.998% (REO, rare earth oxide basis), REacton®

Fornitore: Thermo Fisher Scientific

Synonyms: Diytterbium trioxide, Ytterbium oxide

10919.06 10919.14
10919.06EA 82.9 EUR
10919.06 10919.14
Itterbio (III) ossido ≥99.998% (REO, rare earth oxide basis), REacton®
Itterbio (III) ossido

Purity: 99.998% CAS No.: 1314-37-0 Molecular Formula: Yb2O3 Formula Weight: 394.08 Physical Form: Powder Density: 9.2 Melting Point: 2227° Merck Reference: 14,10105 MDL No.: MFCD00011290

Formula: O₃Yb₂
Peso molecolare: 394.08 g/mol
Punto di fusione: 2227 °C
Densità: 9.2
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00011290
CAS Number: 1314-37-0
EINECS: 215-234-0
Merck Index: 14,10105

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR