Stampa…
Avantor® Seradigm Foetal Bovine Serum (FBS), Ultra-Low IgG, US Origin
Fornitore: VWR Seradigm
10018-826EA 0 EUR
10018-826 10018-828 10836-696 10836-698 10018-830 10018-832
Avantor® Seradigm Foetal Bovine Serum (FBS), Ultra-Low IgG, US Origin
Sieri
Ultra-low IgG foetal bovine serum (FBS) is purified of immunoglobulin (IgG) using a chromatographic process that delivers significantly reduced levels of IgG (≤5 μg/ml).
  • 100% US origin
  • Triple 0,1 μm sterile filtration
  • Preservation of native, growth-promoting factors and minimisation of growth-inhibiting factors
  • Independently tested by third party laboratories
  • Fully traceable
Ultra-low IgG FBS is ideal for cell culture and protein purification applications where naturally occurring levels of IgG are too high.
Certificazioni: International SerumMore Product Information