Stampa…

Acido cloridrico 0.01 M, Titripur®, Supelco®

Fornitore: Merck


M-Clarity™ quality level = MQ100

1.60238.1000
1.60238.1000EA 20.3 EUR
1.60238.1000
Acido cloridrico 0.01 M, Titripur®, Supelco®
Acido cloridrico
Formula: HCl
MDL Number: MFCD00011324
CAS Number: 7647-01-0

Visualizza i codici

Risultati del test di specificazione

Storage Store at +15 °C to +25 °C
Molarity 0,01 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR