Stampa…
HPLC columns, LiChroCART®, Purospher® STAR NH₂
Fornitore: Merck
1.50229.7177 1.50195.7177 1.50234.7177 1.50201.7177 1.50177.7177 1.50179.7177 1.51431.7177 1.50174.7177 1.50175.7177 1.50228.7177 1.50233.7177 1.50236.7177 1.50302.7177 1.51442.7177 1.51443.7177 1.50170.7177 1.51432.7177 1.50173.7177 1.51444.7177 1.51445.7177 1.50171.7177 1.51448.7177 1.50178.7177
1.50229.7177EA 378 EUR
1.50229.7177 1.50195.7177 1.50234.7177 1.50201.7177 1.50177.7177 1.50179.7177 1.51431.7177 1.50174.7177 1.50175.7177 1.50228.7177 1.50233.7177 1.50236.7177 1.50302.7177 1.51442.7177 1.51443.7177 1.50170.7177 1.51432.7177 1.50173.7177 1.51444.7177 1.51445.7177 1.50171.7177 1.51448.7177 1.50178.7177
HPLC columns, LiChroCART®, Purospher® STAR NH₂
Colonne cromatografiche
Stainless steel LiChroCART® cartridges are available with various types of Purospher® sorbents.
These LiChroCART® cartridges require part number 1.51486.0001 manu-CART® cartridge column holder, which can be used to hold one cartridge column with or without a  4×4 mm guard column. LiChroCART® columns 250×10 mm require part number 1.51419.0001 manu-CART® 10. The short LiChroCART® columns can be ordered as a set including the corresponding cartridge holder and one cartridge, or as a pack of three cartridges without cartridge holder. The separate part numbers for the cartridge are asMore Product Information