Stampa…

Polivinil alcool, M.W. 115,000 ≥88%, EMPROVE® EXPERT Ph. Eur., USP, JPE adatto a essere utilizzato come eccipiente, hydrolysed, SAFC®

Fornitore: MERCK PRODUCTION CHEMICALS

Synonyms: PVOH, PVA, Mowiol®

1.41351.1000 1.41351.9029
1.41351.1000EA 520 EUR
1.41351.1000 1.41351.9029
Polivinil alcool, M.W. 115,000 ≥88%, EMPROVE® EXPERT Ph. Eur., USP, JPE adatto a essere utilizzato come eccipiente, hydrolysed, SAFC®
Polivinil alcool
Formula: [-CH₂CH(OH)-]n
MDL Number: MFCD00081922
CAS Number: 9002-89-5

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR