Stampa…

3-(4,5-Dimetiltiazol-2-il)-2,5-difeniltetrazolio bromuro (MTT), Sigma-Aldrich®

Fornitore: Merck
Attenzione


M-Clarity™ quality level = MQ100

1.11714.0001 1.11714.0010
1.11714.0001EA 242 EUR
1.11714.0001 1.11714.0010
3-(4,5-Dimetiltiazol-2-il)-2,5-difeniltetrazolio bromuro (MTT), Sigma-Aldrich®
3-(4,5-Dimetiltiazol-2-il)-2,5-difeniltetrazolio bromuro (MTT)
Formula: C₁₈H₁₆N₅BrS
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00011964
CAS Number: 298-93-1
EINECS: 206-069-5

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR