Stampa…

Acido cloridrico concentrate for dilution to 1 mol/l in soluzione acquosa, Titrisol®, Supelco®

Fornitore: Merck
Attenzione

Synonyms: Hydrochloric acid solution, Muriatic acid

M-Clarity™ quality level = MQ100

1.09970.0001 1.09970.0050
1.09970.0001EA 24.66 EUR
1.09970.0001 1.09970.0050
Acido cloridrico concentrate for dilution to 1 mol/l in soluzione acquosa, Titrisol®, Supelco®
Acido cloridrico
Formula: HCl
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,

Visualizza i codici

Risultati del test di specificazione

Solubility (20 °C) soluble
Density 1.09 g/cm3 (20 °C)
pH  - 1 (H2O, 20 °C)
Hazard Class 8
Hazard Class Description Corrosive Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR