Stampa…
Soluzioni tampone colorate Certipur®, Supelco®
Fornitore: Merck
1.09475.0500 1.09475.9010 1.09476.0500 1.09476.9010 1.09477.0500 1.09477.9010 1.09400.0500 1.09400.9010 1.09400.4000 1.09475.4000 1.09476.4000 1.09477.4000
1.09475.0500EA 44.52 EUR
1.09475.0500 1.09475.9010 1.09476.0500 1.09476.9010 1.09477.0500 1.09477.9010 1.09400.0500 1.09400.9010 1.09400.4000 1.09475.4000 1.09476.4000 1.09477.4000
Soluzioni tampone colorate Certipur®, Supelco®
Soluzioni tampone Tamponi pH
Soluzioni per una calibrazione precisa e il controllo dei pHmetri.
  • Traceable to NIST primary standard and PTB - Uncertainty ±0,015 (except pH 10 with ±0,05)
  • Colour coded solutions for quick identification
  • Larger pack sizes available, the easy to use Titripac® packaging eliminates contamination issues
  • Reference temperature: +20 °C
Measured in an independent accredited calibration laboratory for the pH value measurement (DIN EN ISO/IEC 17025).
Visualizza i codici
Specifiche
M-Clarity quality level MQ300 MQ300
Environmentally Preferable Imballi sostenibili