Stampa…

Buffer solutions, ready to use, referenced at 25 °C, Certipur®

Fornitore: Merck

1.09444.0500 1.09445.0500 1.09445.4000 1.99038.0500 1.99054.0500 1.99054.4000 1.09446.0500 1.99036.0500 1.09406.4000 1.09407.0500 1.09407.4000 1.09408.4000 1.09409.0500 1.09409.4000 1.09442.0500 1.09442.4000 1.09408.0500 1.09441.0500 1.99057.0500 1.99057.4000 1.99050.0500 1.99050.4000 1.09441.4000 1.99041.0500 1.99041.4000
1.09444.0500EA 46.7 EUR
1.09444.0500 1.09445.0500 1.09445.4000 1.99038.0500 1.99054.0500 1.99054.4000 1.09446.0500 1.99036.0500 1.09406.4000 1.09407.0500 1.09407.4000 1.09408.4000 1.09409.0500 1.09409.4000 1.09442.0500 1.09442.4000 1.09408.0500 1.09441.0500 1.99057.0500 1.99057.4000 1.99050.0500 1.99050.4000 1.09441.4000 1.99041.0500 1.99041.4000
Buffer solutions, ready to use, referenced at 25 °C, Certipur®
Soluzioni tampone Tamponi pH

Materials for the precise calibration and monitoring of pH meters.


  • Traceable to NIST primary standard and PTB - deviation max. ±0,01 (except pH 10 with ±0,02)
  • Larger 4 litre pack sizes in easy to use Titripac® packaging which eliminates contamination issues
  • Colour coded buffers at pH values 4,00, 7,00 and 10,00 are available
  • Reference temperature: +25 °C


All buffer solutions are measured and qualified in a DIN EN ISO 17025/IEC accredited laboratory for pH measurement.


Confezione: Plastic bottles (500 ml) or PE/aluminium Titripac® pack (4 litre)

Visualizza i codici

Specifiche

Environmentally Preferable Imballi sostenibili

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR