Stampa…
Detergenti Extran® per macchine lavavetreria (AP), Supelco®
Fornitore: Merck
1.07558.2000 1.07558.9025 1.07559.2500 1.07559.9010 1.07559.9025 1.07561.2500 1.07561.9010 1.07561.9025 1.07563.2000 1.07563.9010 1.07563.9025 1.07570.2000 1.07570.9025 1.07558.9010 1.40006.2500 1.40006.9010 1.40006.9025 1.40006.5000 1.40118.2500 1.40118.5000 1.40118.9010 1.40118.9025 1.07559.5001 1.07561.5001 1.40006.5001 1.40118.5001
1.07558.2000EA 163.5 EUR
1.07558.2000 1.07558.9025 1.07559.2500 1.07559.9010 1.07559.9025 1.07561.2500 1.07561.9010 1.07561.9025 1.07563.2000 1.07563.9010 1.07563.9025 1.07570.2000 1.07570.9025 1.07558.9010 1.40006.2500 1.40006.9010 1.40006.9025 1.40006.5000 1.40118.2500 1.40118.5000 1.40118.9010 1.40118.9025 1.07559.5001 1.07561.5001 1.40006.5001 1.40118.5001
Detergenti Extran® per macchine lavavetreria (AP), Supelco®
Soluzioni detergenti Detergenti e Detersivi per Superfici
Una gamma di detergenti, per macchine lavavetreria, sviluppata per soddisfare le necessità di lavaggio di vetreria e della piccola strumentazione in laboratorio.
  • Prevents carry-over of residues to the next analysis
  • Chlorine-free
These products have been developed with leading instrument manufacturers for their suitability for washing machines. Extran® products have been tested extensively for their effectiveness and low degree of foaming. It is advisable to neutralise alkaline cleansers with an acid rinse subsequent to the cleaning process. AlkalineMore Product Information