Stampa…

n-Pentano, Uvasol® per spettroscopia, Supelco®

Fornitore: Merck
Pericolo


M-Clarity™ quality level = MQ100

1.07179.1000
1.07179.1000EA 229 EUR
1.07179.1000
n-Pentano, Uvasol® per spettroscopia, Supelco®
n-Pentano

Accurate analytic results in UV/Vis and infrared spectroscopy depend on the use of very pure solvents for sample preparation. The Uvasol® solvents range has been specially designed for spectroscopy and other applications requiring solvents of the highest spectral purity. The refinement process allows a greater degree of security in applications and avoids misinterpretation of analytical results caused by traces of UV, IR and fluorescence contamination. Uvasol® solvents offer best UV transmittance. In all specifications the minimum transmittance for five typical wavelengths are identified. Furthermore the transmittance is specified in accordance with Reag. Ph. Eur. and ACS.

Formula: H₃C(CH₂)₃CH₃
Punto di ebollizione: 35…36 °C (1013 hPa)
Punto di fusione: –130 °C
Densità: 0,63 g/cm³ (20 °C)
Flash Pt: –40 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00009498
CAS Number: 109-66-0
EINECS: 203-692-4
UN: 1265
ADR: 3,II
REACH: 01-2119459286-30
Merck Index: 13,07193
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ba2c9c81a4b1f44769627a00ba49b6cc8e63132&vid=f6831eb15ab1602d8c752468dc3ee31cc4ee61c0&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Solubility 0.04 g/L (25 °C)
Melting Point -130 °C
Molar Mass 72.15 g/mol
Boiling Point 36 °C (1013 hPa)
Vapor Pressure 570 hPa (20 °C)
Flash Point -48 °C (refers to pure substance)
Refractive Index 1.358
Explosion Limit 1.4 - 8%(V)
Density 0.63 g/cm³ (20 °C)
pH (H₂O) no data available
Ignition Point 285 °C
Hazard Class 3
Hazard Class Description Flammable liquid

Specifiche

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR