Stampa…

Mercurio (II) ioduro, rosso Reag. Ph. Eur., ACS per analisi, Supelco®

Fornitore: Merck
Pericolo

Synonyms: Mercury diiodide, Mercuric iodide

M-Clarity™ quality level = MQ300

1.04428.0050 1.04428.0250
1.04428.0050EA 138 EUR
1.04428.0050 1.04428.0250
Mercurio (II) ioduro, rosso Reag. Ph. Eur., ACS per analisi, Supelco®
Mercurio (II) ioduro
Formula: HgI₂
Punto di ebollizione: 354 °C (1013 hPa)
Punto di fusione: 259 °C
Densità: 6,36 g/cm³ (20 °C)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00011044
CAS Number: 7774-29-0
EINECS: 231-873-8
UN: 1638
ADR: 6.1,II
Merck Index: 13,05905

Visualizza i codici

Risultati del test di specificazione

Assay (argentometric, dried sust.) ≥ 99.0 %
Solubility in Potassium iodide passes test
Soluble mercury salts (as Hg) ≤ 0.05 %
Foreign heavy metals (as Pb) ≤ 0.001 %
Fe (Iron) ≤ 0.001 %
Mercury (I) (as Hg) ≤ 0.1 %
Substances not reducing by formic acid ≤ 0.02 %
Corresponds to ACS,Reag. Ph Eur 6 - 7 (50 g/l, H2O, 20 °C) (slurry)
Hazard Class 6.1
Hazard Class Description Toxic materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR