Stampa…

n-Esano, SupraSolv® GC-ECD/FID, Supelco®

Fornitore: Merck
Pericolo

Synonyms: Hexyl hydride, Hexanen, Hexane

MQ100

1.04371.1000 1.04371.2500 1.04371.4000 1.04371.9030
1.04371.1000EA 45.9 EUR
1.04371.1000 1.04371.2500 1.04371.4000 1.04371.9030
n-Esano, SupraSolv® GC-ECD/FID, Supelco®
n-Esano

SupraSolv® ECD and FID is specially developed for gas chromatography in combination with ECD (Electron Capture Detetor) and FID (Flame Ionization detector). SupraSolv® solvents offer the largest specified retention time range, a clear baseline and a minimal signal-to-noise ratio leading to reliable and reproducible analysis results.

Formula: H₃C(CH₂)₄CH₃
Peso molecolare: 86.18 g/mol
Punto di ebollizione: 69
Punto di fusione: –94.3 °C
Densità: 0.659 g/cm³ (20 °C)
Flash Pt: –22 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00009520
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3 II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Promozione a tempo limitato

Questi articoli sono attualmente in promozione

Visualizza i codici

Risultati del test di specificazione

Purity (GC) ≥98.0 %
Sum of hexane isomers + methylcyclopentane (GC) ≥99.8 %
Identity (IR) conforms
Evaporation residue ≤3.0 mg/l
Water ≤0.01 %
Color ≤ 10 Hazen
GC/ECD (retention range 1,2,4-tri­chlorobenzene to decachlorobiphenyle, individual signals (lindane standard)) ≤ 3 pg/ml
GC/FID (retention range n-undecane to n-tetracontane,individual signals (n-tetradecane standard)) ≤ 3 ng/ml
suitable for residue analysis 0.66 g/cm3 (20 °C)
pH (H2O) not applicable
Ignition Point 240 °C
Viscosity kinematic 0.50 mm2/s (20 °C)

Specifiche

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR