Stampa…
Glass beads
Fornitore: Merck
1.04014.0500 1.04015.0500 1.04016.0500 1.04018.0500 1.04017.0500
1.04014.0500EA 67.7 EUR
1.04014.0500 1.04015.0500 1.04016.0500 1.04018.0500 1.04017.0500
Glass beads
Sferette Sferette in vetro
Soda-lime glass. Glass beads for use in purification and sample preparation.
Glass beads of size 2 to 6 mm can be used in a wide range of ways in laboratories and technical facilities: For illing distillation columns (the size of the glass beads determines the distillation speed and separation); As agitator and mixing beads e.g. when mixing solutions which tend to settle or substances which are difficult to dissolve; For the prevention of superheating during distillation, especially in highly pure solvents or acids; In the concentration or vaporization of solutions (prevents change inMore Product Information