Stampa…

4-Clorofenolo soluzione 725 mg/ml DIN EN ISO 9562 for AOX determination, Supelco®

Fornitore: Merck


M-Clarity™ quality level = MQ100

1.03085.0100
1.03085.0100EA 101 EUR
1.03085.0100
4-Clorofenolo soluzione 725 mg/ml DIN EN ISO 9562 for AOX determination, Supelco®
4-Clorofenolo

4-Chlorophenol solution for AOX determination acc. to DIN EN ISO 9562.

Formula: C₆H₅ClO
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00002318
CAS Number: 106-48-9
ADR: 6.1,III

Visualizza i codici

Risultati del test di specificazione

Density 1.00 g/cm³ (20 °C)
pH 5 (H₂O, 20 °C)
Storage Class 10 - 13 Other liquids and solids
WGK NWG not water endangering
Storage Store at +15°C to +25°C.
Purity ≥99.0 %
Identity Passes test
Halides ≤100 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR