Stampa…

Etanolo 96% (v/v), EMPROVE® EXPERT Ph. Eur., BP, ChP, extra puro, SAFC®

Fornitore: MERCK PRODUCTION CHEMICALS
Pericolo

Synonyms: Ethyl alcohol

1.00971.1000 1.00971.2500 1.00971.6010 1.00971.6025 1.00971.2500DP 1.00971.1000DP 1.00971.1000_T 1.00971.2500_T 1.00971.6010_T 1.00971.6025_T 1.00971.6190_T 1.00971.6010DP 1.00971.6025DP
1.00971.1000EA 124 EUR
1.00971.1000 1.00971.2500 1.00971.6010 1.00971.6025 MERP1.00971.2500DP MERP1.00971.1000DP MERP1.00971.1000_T MERP1.00971.2500_T MERP1.00971.6010_T MERP1.00971.6025_T MERP1.00971.6190_T MERP1.00971.6010DP 1.00971.6025DP
Etanolo 96% (v/v), EMPROVE® EXPERT Ph. Eur., BP, ChP, extra puro, SAFC®
Etanolo
Formula: C₂H₅OH
Peso molecolare: 46.07 g/mol
Punto di ebollizione: 78 °C (1013 hPa)
Punto di fusione: –114 °C
Densità: 0.805…0.812 g/cm3 (20 °C)
Flash Pt: 17 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00003568
CAS Number: 64-17-5
EINECS: 200-578-6
UN: 1170
ADR: 3 II
Merck Index: 12,03806

Visualizza i codici

Risultati del test di specificazione

Assay (m/m) 92.6 - 95.2 %
Assay (V/V) 95.1 - 96.9 %
Identity (IR) conforms
Identity 1 (ChP) conforms
Appearance conforms
Clarity and color of solution (ChP) conforms
Acidity or alkalinity ≤ 30 ppm
Acidity or alkalinity (ChP) conforms
Density (d 20 °C/20 °C) 0.805 - 0.812
Density (d 20 °C/20 °C) (ChP) ≤ 0.8129
Absorbance (at 240 nm) ≤ 0.40
Absorbance (between 250 nm and 260 nm) ≤ 0.30
Absorbance (between 270 nm and 340 nm) ≤ 0.10
The spectrum shows a steadily descending curve with no observable peaks or shoulders conforms
Absorbance (ChP) (at 240 nm) ≤ 0.08
Absorbance (ChP) (between 250 nm and 260 nm) ≤ 0.06
Absorbance (ChP) (between 270 nm and 340 nm) ≤ 0.02
Volatile impurities (GC) (Acetaldehyde and Acetal) ≤ 10 ppm
Volatile impurities (GC) (Benzene) ≤ 2 ppm
Volatile impurities (GC) (Methanol) ≤ 100 ppm
Volatile impurities (GC) (Total of other impurities) ≤ 300 ppm
Other residual solvents (ICH Q3C) excluded by production process
Evaporation residue ≤ 25 mg/l
Non-volatile matter ≤ 25 mg/l
Bacterial endotoxins ≤ 2.5
Staphylococcus aureus (absent in 1 g) passes test
Parameter with periodic testing
Total aerobic microbial count (TAMC) ≤ 100 CFU/ml
Total combined yeasts/moulds count (TYMC) ≤ 10 CFU/ml
Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).
Conforms to Ph Eur, BP, ChP.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR