Stampa…
Kit analitici, sodio in cuvetta in soluzioni nutritive per fertilizzazione per Spectroquant®, Supelco®
Fornitore: Merck
1.00885.0001
1.00885.0001EA 211 EUR
1.00885.0001
Kit analitici, sodio in cuvetta in soluzioni nutritive per fertilizzazione per Spectroquant®, Supelco®
Kit per analisi ambientali
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request).
  • Very user friendly, validated methods
  • All the necessary reagents for the analysis are contained in the cells for most the test parameters
  • Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
  • The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times.
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.