Stampa…

Acido acetico ≥96%, EMPROVE® ESSENTIAL, SAFC®

Fornitore: MERCK PRODUCTION CHEMICALS
Pericolo

1.00058.2500 1.00058.9025
1.00058.2500EA 189 EUR
1.00058.2500 1.00058.9025
Acido acetico ≥96%, EMPROVE® ESSENTIAL, SAFC®
Acido acetico
Formula: H₃CCOOH
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00036152
CAS Number: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8 (3),II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR