Stampa…

Acido tricloroacetico (glaciale) ≥99.0% (by acidimetry), cristalli, BAKER ANALYZED® ACS, J.T.Baker®

Fornitore: Avantor
Pericolo

0414-04EA 162.34 EUR
0414-04
Acido tricloroacetico (glaciale) ≥99.0% (by acidimetry), cristalli, BAKER ANALYZED® ACS, J.T.Baker®
Acido tricloroacetico (glaciale)

Crystals. Lot analysis on label.


  • Lot analysis on label

Formula: Cl₃CCOOH
Punto di ebollizione: 196 °C (1013 hPa)
Punto di fusione: 54…56 °C
Densità: 1,63 g/cm³ (20 °C)
Flash Pt: 113 °C (coppa chiusa)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00004177
CAS Number: 76-03-9
EINECS: 200-927-2
Merck Index: 13,09700
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae34130684af3f73eccad44eac686e325855232d&vid=b600bf2ba67ce275263e6172675e131d3f263837&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Meets ACS Reagent Chemical Requirements ≥99.0%
For Laboratory,Research,or Manufacturing Use Passes Test
IMPORTANT: Store or transport at or below 30°C (86°F). ≤0.01%
Assay (Cl₃CCOOH) (by acidimetry) ≥99.0 %
Clarity of Solution Passes Test
Insoluble Matter ≤0.01 %
Residue after Ignition ≤0.03 %
Chloride (Cl) ≤0.002 %
Nitrate (NO₃) ≤0.002 %
ACS - Phosphate (PO₄) ≤5 ppm
Sulfate (SO₄) ≤0.02 %
Heavy Metals (as Pb) ≤0.002 %
Trace Impurities - Iron (Fe) ≤0.001 %
Substances Darkened by H₂SO₄ Passes Test
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR