Stampa…

Acido o-fosforico 33% (w/w), Biotech Reagent for biotechnology, J.T.Baker®

Fornitore: Avantor
Pericolo

Synonyms: Acido fosforico

filtrato a 0,2 µm

0334-07EA 920.66 EUR
0334-07 0334-09 0334-27
Acido o-fosforico 33% (w/w), Biotech Reagent for biotechnology, J.T.Baker®
Acido o-fosforico

Made from USP purified water and phosphoric acid, NF (multicompendial) which meets BP and Ph. Eur. chemical specifications.

Formula: H₃PO₄
Punto di fusione: 169…170 °C
Densità: 1,29 g/cm³ (20 °C)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00011340
CAS Number: 7664-38-2
EINECS: 200-334-9
UN: 1805
ADR: 8,III
Merck Index: 13,08966

Visualizza i codici

Risultati del test di specificazione

GMP Manufactured Product 32.0 - 34.0%
CAUTION: For Manufacturing, processing or repackaging Passes Test
Assay (H₃PO₄) 32.0 - 34.0 %
Alkali Phosphates Passes Test
ACS - Heavy Metals (as Pb) ≤5 ppm
Nitrate (NO₃) Passes Test
Sulfate (SO₄) Passes Test
Identification Passes Test
Phosphorous or Hypophosphorous Acid Passes Test
Arsenic (As) ≤0.200 ppm
Appearance of Solution Passes Test
Chloride (Cl) ≤6 ppm
Made from U.S.P. Purified Water and Phosphoric Acid, N.F. (Multicompendial) which meets B.P. and Ph.Eur. Chemical Specifications.
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.
Filtered through a 0.2 micron filter.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR