Stampa…

Acido o-fosforico ≥85.0-88.0% NF, FCC, J.T.Baker®

Fornitore: Avantor
Pericolo


Management of Change (MOC) category = R

0262-07EA 2086.36 EUR
0262-07 0262-02 0262-05 0262-01 0262-03 0262-06 0262-04 0262-08 0262-15
Acido o-fosforico ≥85.0-88.0% NF, FCC, J.T.Baker®
Acido o-fosforico
Formula: H₃PO₄
Punto di fusione: 28 °C
Densità: 1,69 g/cm³
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00011340
CAS Number: 7664-38-2
EINECS: 231-633-2
UN: 1805
ADR: 8,III

Visualizza i codici

Risultati del test di specificazione

Food GMP Manufactured Product 85.0 - 88.0%
GMP Manufactured Product Passes Test
Meets N.F. Requirements Passes Test
Meets F.C.C. Requirements Passes Test
CAUTION: For Manufacturing, processing or repackaging Passes Test
Bulk Food Chemical Passes Test
Bulk Pharmaceutical Chemical 85.0 - 88.0%
NF - Assay (H₃PO₄) 85.0 - 88.0 %
NF - Identification Passes Test
NF - Nitrate (NO₃) Passes Test
NF - Phosphorous or Hypophosphorous Acid Passes Test
NF - Sulfate (SO₄) Passes Test
NF - Alkali Phosphates Passes Test
FCC - Assay (H₃PO₄) 85.0 - 88.0 %
FCC - Identification Passes Test
FCC - Arsenic (As) ≤3 mg/kg
FCC - Cadmium (Cd) ≤3 mg/kg
FCC - Lead (Pb) ≤3 mg/kg
FCC - Fluoride (F) ≤10 mg/kg
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232) - Information on elemental impurities for
this product is available on the associated Product Regulatory Data
Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR