Stampa…

Acido borico ≥99.5% (by acidimetry), granular, BAKER ANALYZED® ACS, J.T.Baker®

Fornitore: Avantor
Pericolo

0084-01EA 94.51 EUR
0084-01 0084-07 0084-08
Acido borico ≥99.5% (by acidimetry), granular, BAKER ANALYZED® ACS, J.T.Baker®
Acido borico

Meets Reagent Specifications for testing USP/NF monographs.

Formula: B(OH)₃
Punto di ebollizione: 185 °C (1013 hPa)
Punto di fusione: 160 °C
Densità: 1,44 g/cm³ (25 °C)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00011337
CAS Number: 10043-35-3
EINECS: 233-139-2
REACH: 01-2119486683-25
Merck Index: 13,01326
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=098dd700154daaf6338ebb270902bb2bc704f27d&vid=6eec7d529090dd4c4b966ed8cd8d057153c5a47b&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Meets ACS Reagent Chemical Requirements ≥99.5%
For Laboratory,Research,or Manufacturing Use ≤0.005%
Assay (H₃BO₃) (by acidimetry) ≥ 99.5 %
Insoluble in Methanol ≤ 0.005 %
Nonvolatile with Methanol ≤ 0.05 %
Chloride (Cl) ≤ 0.001 %
Phosphate (PO₄) ≤ 0.001 %
Sulfate (SO₄) ≤ 0.005 %
Calcium (Ca) ≤ 0.005 %
Heavy Metals (as Pb) ≤ 0.001 %
Trace Impurities - Iron (Fe) ≤ 0.001 %
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR