Stampa…

3,6-Dimethylcarbazole ≥98.0% (by GC)

Fornitore: TCI
Attenzione

Synonyms: 3,6-Dimethyl-9H-carbazole

D4512-1G D4512-5G
TCIAD4512-1GEA 219 EUR
TCIAD4512-1G TCIAD4512-5G
3,6-Dimethylcarbazole ≥98.0% (by GC)
3,6-Dimethylcarbazole
Formula: C₁₄H₁₃N
Peso molecolare: 195,27 g/mol
MDL Number: MFCD00814049
CAS Number: 5599-50-8

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 220.0 to 225.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR