Stampa…

Anti-EBV-Induced Gene 3 Goat Polyclonal Antibody

Fornitore: US Biological
E3440-50A
USBIE3440-50AEA 473 EUR
USBIE3440-50A
Anti-EBV-Induced Gene 3 Goat Polyclonal Antibody
Anticorpi
Recommended dilution: ELISA: 1:32000

Type: Primary
Antigen: EBV-Induced Gene 3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR