Stampa…
MicroPlates, 96-well, Whatman™ UNIFILTER®
Fornitore: Whatman products (Cytiva)
7700-2801 7700-2803 7700-2808 7700-2810 7700-2804
734-2522EA 1540 EUR
734-2522 734-2523 734-2524 734-2525 738-0200
MicroPlates, 96-well, Whatman™ UNIFILTER®
Micropiastre Piastre filtranti
UNIFILTER 96-well microplate, 800 µl, long drip director.
  • No cross talk: Patented integral filter design prevents well-to-well cross contamination
  • Economical to use: Wide range of well volume options ensures efficient use of materials
  • Better control: Choice of filter media allows control of the flow rates and retention characteristics
  • Versatile: A broad range of filtration media is available, including glass fiber, polypropylene, cellulose nitrate, cellulose acetate, nylon, and ion exchange cellulose
The 800 µl UNIFILTER microplate is most typicallyMore Product Information